Få mere at vide
Cimetidine EP Impurity A, Mikromol™
Artikelnummer. 30050658
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050658 25 mg
30050372 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050658 Leverandør Mikromol Leverandørnr. MM0020.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Cimetidine EP Impurity A
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Cyano-2-methyl-1-[2-[((5-methyl-1H-imidazol-4-yl)methyl)sulphanyl]ethyl]isothiourea
CAS 52378-40-2
Analyt- eller komponentnavne 3-Cyano-2-methyl-1-[2-[((5-methyl-1H-imidazol-4-yl)methyl)sulphanyl]ethyl]isothiourea
Forsendelsestilstand Room Temperature
Molekylær formel C10 H15 N5 S2
Certificeringer/compliance ISO 17025
Synonym 3-Cyano-2-methyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isothiourea, Cimetidine Hydrochloride Imp. A (EP), Cimetidine Imp. A (EP), 3-Cyano-2-methyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isothiourea, Cimetidine Hydrochloride Impurity A, Cimetidine Impurity A, Cimetidine Impurity A
InChI formel InChI=1S/C10H15N5S2/c1-8-9(15-7-14-8)5-17-4-3-12-10(16-2)13-6-11/h7H,3-5H2,1-2H3,(H,12,13)(H,14,15)
SMIL CSC(=NC#N)NCCSCc1nc[nH]c1C
IUPAC navn methyl N'-cyano-N-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethyl]carbamimidothioate
Molekylvægt (g/mol) 269.39
Mængde 25 mg
Formel vægt 269.0769 g/mol