Få mere at vide
Cimetidine Amide Dihydrochloride, TRC
Artikelnummer. 30353331
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30353331 5 mg
30350694 25 mg
30352833 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30353331 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4416605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 52568-80-6
Molekylær formel C10 H18 N6 O S . 2 Cl H
Molekylvægt (g/mol) 343.28
Synonym 1-[(Methylamino)[[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]amino]methylene]urea Dihydrochloride, Cimetidine Hydrochloride Imp. C (EP) as Dihydrochloride, Cimetidine Amide Dihydrochloride, Cimetidine Imp. C (EP) as Dihydrochloride
IUPAC navn [methylamino-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethylamino]methylidene]urea;dihydrochloride
SMIL Cl.Cl.CNC(=NC(=O)N)NCCSCc1nc[nH]c1C
Tekniske data
Kemisk navn eller materiale Cimetidine Amide Dihydrochloride
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C10H18N6OS.2ClH/c1-7-8(15-6-14-7)5-18-4-3-13-10(12-2)16-9(11)17;;/h6H,3-5H2,1-2H3,(H,14,15)(H4,11,12,13,16,17);2*1H
Formel vægt 342.0796
Procent renhed >95%