Få mere at vide
Cimbuterol-d9, TRC
Artikelnummer. 30363775
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30363775 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30363775 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4416221MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cimbuterol D9 (tert-butyl D9)
CAS 1246819-04-4
Renhedsgrad noter HPLC
Molekylær formel C13 2H9 H10 N3 O
Mængde 1 mg
Synonym 2-Amino-5-[2-[[1,1-di(methyl-d3)ethyl-2,2,2-d3]amino]-1-hydroxyethyl]benzonitrile, D9-Cimbuterol, Benzonitrile, 2-amino-5-[2-[[1,1-di(methyl-d3)ethyl-2,2,2-d3]amino]-1-hydroxyethyl]-
InChI formel InChI=1S/C13H19N3O/c1-13(2,3)16-8-12(17)9-4-5-11(15)10(6-9)7-14/h4-6,12,16-17H,8,15H2,1-3H3/i1D3,2D3,3D3
SMIL [2H]C([2H])([2H])C(NCC(O)c1ccc(N)c(c1)C#N)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 2-amino-5-[2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]-1-hydroxyethyl]benzonitrile
Molekylvægt (g/mol) 242.36
Formel vægt 242.2093
Procent renhed >95