Få mere at vide
Cilostazol-d4, TRC
Artikelnummer. 30361541
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361541 1 mg
30368688 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361541 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4415031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1215541-47-1
Molekylær formel C20H23D4N5O2
Molekylvægt (g/mol) 373.49
Synonym 6-[4-(1-Cyclohexyl-d4-1H-tetrazol-5-yl)butoxy]-3,4-dihydro-2(1H)-quinolinone, OPC-13013-d4, Pletal-d4
IUPAC navn 6-[4-[1-(2,2,6,6-tetradeuteriocyclohexyl)tetrazol-5-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
SMIL O=C(N1)CCC2=C1C=CC(OCCCCC3=NN=NN3C4C([2H])([2H])CCCC4([2H])[2H])=C2
Tekniske data
Kemisk navn eller materiale Cilostazol-d4
Mængde 1 mg
InChI formel InChI=1S/C20H27N5O2/c26-20-12-9-15-14-17(10-11-18(15)21-20)27-13-5-4-8-19-22-23-24-25(19)16-6-2-1-3-7-16/h10-11,14,16H,1-9,12-13H2,(H,21,26)/i6D2,7D2