Få mere at vide
Cilazapril Monohydrate, Mikromol™
Artikelnummer. 17911886
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
17911886 250 mg
1 options
Artikelnummer. 17911886 Leverandør Mikromol Leverandørnr. MM3519.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Cilazapril Monohydrate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cilazapril Monohydrate
CAS 92077-78-6
Analyt- eller komponentnavne Cilazapril Monohydrate
Forsendelsestilstand Room Temperature
Molekylær formel C22 H31 N3 O5 . H2 O
Certificeringer/compliance ISO 17034
Synonym (1S,9S)-9-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]octahydro-10-oxo-6H-pyridazino[1,2-a][1,2]diazepine-1-carboxylic Acid, Ro 31-2848, Vascace, Yipingshu, Cilazapril, (1S,9S)-9-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-10-oxooctahydro-6H-pyridazino[1,2-a][1,2]diazepine-1-carboxylic acid monohydrate, 6H-Pyridazino[1,2-a][1,2]diazepine-1-carboxylic acid, 9-[[(1S)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]octahydro-10-oxo-, hydrate (1:1), (1S,9S)-, 6H-Pyridazino[1,2-a][1,2]diazepine-1-carboxylic acid, 9-[[(1S)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]octahydro-10-oxo-, monohydrate, (1S,9S)- (9CI), 6H-Pyridazino[1,2-a][1,2]diazepine-1-carboxylic acid, 9-[[1-(ethoxycarbonyl)-3-phenylpropyl]amino]octahydro-10-oxo-, monohydrate, [1S-[1alpha,9alpha(R*)]]-, Cilazapril monohydrate
InChI formel InChI=1S/C22H31N3O5.H2O/c1-2-30-22(29)18(13-12-16-8-4-3-5-9-16)23-17-10-6-14-24-15-7-11-19(21(27)28)25(24)20(17)26;/h3-5,8-9,17-19,23H,2,6-7,10-15H2,1H3,(H,27,28);1H2/t17-,18-,19-;/m0./s1
SMIL CCOC(=O)C(CCC1=CC=CC=C1)NC2CCCN3CCCC(N3C2=O)C(=O)O.O
IUPAC navn (4S,7S)-7-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]-6-oxo-1,2,3,4,7,8,9,10-octahydropyridazino[1,2-a]diazepine-4-carboxylic acid;hydrate
Molekylvægt (g/mol) 417.5