Få mere at vide
Cilastatin-15N-d3, TRC
Artikelnummer. 30351184
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30351184 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30351184 Leverandør Toronto Research Chemicals Leverandørnr. TRCC44110110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cilastatin-15N-d3
CAS 2738376-83-3
Molekylær formel C16 D3 H23 15N N O5 S
Mængde 10 mg
InChI formel InChI=1S/C16H26N2O5S/c1-16(2)8-10(16)13(19)18-12(15(22)23)6-4-3-5-7-24-9-11(17)14(20)21/h6,10-11H,3-5,7-9,17H2,1-2H3,(H,18,19)(H,20,21)(H,22,23)/b12-6-/t10-,11+/m1/s1/i9D2,11D,17+1
SMIL [2H]C([2H])(SCCCC\C=C(/NC(=O)[C@H]1CC1(C)C)\C(=O)O)[C@]([2H])([15NH2])C(=O)O
IUPAC navn (Z)-7-[(2R)-2-(15N)azanyl-2-carboxy-1,1,2-trideuterioethyl]sulfanyl-2-[[(1S)-2,2-dimethylcyclopropanecarbonyl]amino]hept-2-enoic acid
Molekylvægt (g/mol) 362.465
Formel vægt 362.172