missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals CIDEA Antibody (4B9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00001149-M01
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
CIDEA Monoclonal antibody specifically detects CIDEA in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Specifica
CIDEA | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
cell death activator CIDE-A, cell death-inducing DFFA-like effector aCIDE-A | |
CIDEA (AAH31896, 1 a.a. ~ 253 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MRGDRASGGPGNHNGSWAREGPRLGPSWKRGLWSPRGGPNRPAEPSRPLTFMGSQTKRVLFTPLMHPARPFRVSNHDRSSRRGVMASSLQELISKTLDALVIATGLVTLVLEEDGTVVDTEEFFQTLGDNTHFMILEKGQKWMPGSQHVPTCSPPKRSGIARVTFDLYRLNPKDFIGCLNVKATMYEMYSVSYDIRCTGLKGLLRSLLRFLSYSAQVTGQFLIYLGTYMLRVLDDKEERPSLRSQAKGRFTCG | |
0.1 mg | |
Apoptosis | |
1149 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
4B9 | |
Western Blot 1:500, ELISA, Immunohistochemistry, Immunoprecipitation | |
AAH31896 | |
Mouse | |
IgG purified | |
RUO | |
Primary | |
Human | |
Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto