Få mere at vide
CID-2858522, MedChemExpress
Artikelnummer. 16496598
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16496598 10 mg
16479167 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16496598 Leverandør Medchem Express Leverandørnr. HY15530
MedChemExpress CID-2858522 is a highly potent and selective antigen receptor-mediated NF-κB activation inhibitor with an IC50 of 70 nM.
CID-2858522 is a highly potent and selective antigen receptor-mediated NF-κB activation inhibitor with an IC50 of 70 nM.
Tekniske data
Kemisk navn eller materiale CID-2858522
CAS 758679-97-9
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C28H39N3O3
Mængde 10 mg
Opløselighedsinformation DMSO : 25 mg/mL (53.69 mM; Need ultrasonic)
SMIL CC1=C(C)C=C2N=C(NCCCO)N(CC(C3=CC(C(C)(C)C)=C(O)C(C(C)(C)C)=C3)=O)C2=C1
Molekylvægt (g/mol) 465.63
Formel vægt 465.63
Procent renhed 95.52%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death