missing translation for 'onlineSavingsMsg'
Få mere at vide

CID 16020046, MedChemExpress

Artikelnummer. 16467138
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
Pakningsstørrelse:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16467138 10 mg -
16453167 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16467138 Leverandør Medchem Express Leverandørnr. HY16697

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress CID 16020046 is a potent and selective GPR55 antagonist and inhibits GPR55 constitutive activity with an IC50 of 0.15 μM. CID 16020046 inhibits GPR55-mediated Ca2+ signaling and GPR55-mediated ERK1/2 phosphorylation. CID 16020046 reduces wound healing in endothelial cells and is involved in the regulation of platelet function.

CID 16020046 is a potent and selective GPR55 antagonist and inhibits GPR55 constitutive activity with an IC50 of 0.15 μM. CID 16020046 inhibits GPR55-mediated Ca2+ signaling and GPR55-mediated ERK1/2 phosphorylation. CID 16020046 reduces wound healing in endothelial cells and is involved in the regulation of platelet function.

Especificaciones

Kemisk navn eller materiale CID 16020046
CAS 834903-43-4
Farve White
Renhedsgrad noter Research
Molekylær formel C25H19N3O4
Mængde 10 mg
Opløselighedsinformation DMSO : 100 mg/mL (235.05 mM; Need ultrasonic)
SMIL O=C(O)C1=CC=C(N(C2C3=CC=CC(O)=C3)C(C4=C2C(C5=CC=C(C)C=C5)=NN4)=O)C=C1
Molekylvægt (g/mol) 425.44
Formel vægt 425.44
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease
Mostrar más Mostrar menos

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.