Få mere at vide
Tocris Bioscience™ CID 1375606
Beskrivelse
CID 1375606 is a GPR27 agonist. Promotes recruitment of β-arrestin 2 to chimeric GPR27V2 and wild-type GPR27 in a firefly luciferase complementation assay (pEC50 values are 6.34 and 5.1, respectively).
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2,4-Dichloro-N-(phenylcarbamoyl)phenylbenzamide |
| CAS | 313493-80-0 |
| Mængde | 5 mg |
| Molekylær formel | C20H14Cl2N2O2 |
| Renhed | 98% |