Få mere at vide
Ciclopirox, Tocris Bioscience™
GREENER_CHOICE
Pan-histone demethylase inhibitor
Brand:  Tocris Bioscience 6384/10
Encompass
Produktkode 15973271
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Ciclopirox is a pan-histone demethylase inhibitor. Exhibits no significant effect on histone deacetylases. Inhibits MYC-signaling and proliferation of neuroblastoma cells. Promotes Cdc25A-degradation in cancer cells. Antifungal.
Tekniske data
Tekniske data
6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridinone
29342-05-0
>99%
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner