Få mere at vide
Ciclopirox, Tocris Bioscience™
Pan-histone demethylase inhibitor
Brand: Tocris Bioscience 6384/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Ciclopirox is a pan-histone demethylase inhibitor. Exhibits no significant effect on histone deacetylases. Inhibits MYC-signaling and proliferation of neuroblastoma cells. Promotes Cdc25A-degradation in cancer cells. Antifungal.Tekniske data
| 6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridinone | |
| 29342-05-0 | |
| >99% | |
| 50 mg |