Få mere at vide
Ciclopirox Olamine EP Impurity A, Mikromol™
Artikelnummer. 30051877
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051877 25 mg
17963506 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051877 Leverandør Mikromol Leverandørnr. MM0418.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ciclopirox Olamine EP Impurity A
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (RS)-2-(3-Cyclohexyl-5-methyl-4,5-dihydroisoxazol-5-yl)acetic Acid
CAS 1823487-53-1
Analyt- eller komponentnavne (RS)-2-(3-Cyclohexyl-5-methyl-4,5-dihydroisoxazol-5-yl)acetic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C12 H19 N O3
Certificeringer/compliance ISO 17025
Synonym [(5RS)-3-Cyclohexyl-5-methyl-4,5-dihydro-1,2-oxazol-5-yl]acetic Acid, Ciclopirox Olamine Imp. A (EP), Ciclopirox USP Related Compound A, Ciclopirox USP RC A, (3-Cyclohexyl-4,5-dihydro-5-methyl-5-isoxazolyl acetic acid), Ciclopirox Related Compound A (USP), Ciclopirox Imp. A (EP), 3-cyclohexyl-4,5-dihydro-5-methyl-5-isoxazolyl acetic acid, USP Ciclopirox Related Compound A, Ciclopirox Related Compound A, Ciclopirox Olamine Impurity A, Ciclopirox Impurity A
InChI formel InChI=1S/C12H19NO3/c1-12(8-11(14)15)7-10(13-16-12)9-5-3-2-4-6-9/h9H,2-8H2,1H3,(H,14,15)
SMIL CC1(CC(=O)O)CC(=NO1)C2CCCCC2
IUPAC navn 2-(3-cyclohexyl-5-methyl-4H-1,2-oxazol-5-yl)acetic acid
Molekylvægt (g/mol) 225.28
Mængde 25 mg
Formel vægt 225.1365 g/mol