Få mere at vide
Ciclopirox Beta-D-Glucuronide, TRC
Artikelnummer. 30359632
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359632 1 mg
30367788 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30359632 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4328101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 79419-54-8
Molekylær formel C18 H25 N O8
Molekylvægt (g/mol) 383.39
IUPAC navn (2S,3S,4S,5R,6S)-6-(2-cyclohexyl-4-methyl-6-oxopyridin-1-yl)oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMIL CC1=CC(=O)N(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)C(=C1)C3CCCCC3
Tekniske data
Kemisk navn eller materiale Ciclopirox beta-D-Glucuronide
Mængde 1 mg
InChI formel InChI=1S/C18H25NO8/c1-9-7-11(10-5-3-2-4-6-10)19(12(20)8-9)27-18-15(23)13(21)14(22)16(26-18)17(24)25/h7-8,10,13-16,18,21-23H,2-6H2,1H3,(H,24,25)/t13-,14-,15+,16-,18-/m0/s1
Formel vægt 383.158