Få mere at vide
Cicletanine-d4 Hydrochloride, TRC
Artikelnummer. 30368254
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30368254 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30368254 Leverandør Toronto Research Chemicals Leverandørnr. TRCC43270210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cicletanine-d4 Hydrochloride
CAS 1189491-41-5
Molekylær formel C14H9D4Cl2NO2
Mængde 10 mg
Synonym 3-(4-Chlorophenyl-d4)-1,3-dihydro-6-methyl-furo[3,4-c]pyridin-7-ol Hydrochloride, BN 1270-d4, Coverine-d4, Justar-d4, Secletan-d4, Tenstaten-d4
InChI formel InChI=1S/C14H12ClNO2.ClH/c1-8-13(17)12-7-18-14(11(12)6-16-8)9-2-4-10(15)5-3-9;/h2-6,14,17H,7H2,1H3;1H/i2D,3D,4D,5D;
SMIL Cl.[2H]c1c([2H])c(C2OCc3c(O)c(C)ncc23)c([2H])c([2H])c1Cl
IUPAC navn 3-(4-chloro-2,3,5,6-tetradeuteriophenyl)-6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol;hydrochloride
Molekylvægt (g/mol) 302.19