Få mere at vide
Ciclesonide-d7, TRC
Artikelnummer. 30360132
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30360132 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30360132 Leverandør Toronto Research Chemicals Leverandørnr. TRCC43269310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ciclesonide-d7
CAS 1225382-70-6
Molekylær formel C32H37D7O7
Mængde 10 mg
Synonym (11β,16α)-16,17-[[(R)-Cyclohexyl-methylene]bis(oxy)]-11-hydroxy-21-(2-methyl-1-oxopropoxy-d7)-pregna-1,4-diene-3,20-dione, Alvesco-d7, Omnaris-d7, RPR 251526-d7
InChI formel InChI=1S/C32H44O7/c1-18(2)28(36)37-17-25(35)32-26(38-29(39-32)19-8-6-5-7-9-19)15-23-22-11-10-20-14-21(33)12-13-30(20,3)27(22)24(34)16-31(23,32)4/h12-14,18-19,22-24,26-27,29,34H,5-11,15-17H2,1-4H3/t22-,23-,24-,26+,27+,29+,30-,31-,32+/m0/s1/i1D3,2D3,18D
SMIL CC(C)C(=O)OCC(=O)C12C(CC3C1(CC(C4C3CCC5=CC(=O)C=CC45C)O)C)OC(O2)C6CCCCC6
IUPAC navn [2-[(1S,2S,4R,6R,8S,9S,11S,12S,13R)-6-cyclohexyl-11-hydroxy-9,13-dimethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-8-yl]-2-oxoethyl] 2,3,3,3-tetradeuterio-2-(trideuteriomethyl)propanoate
Molekylvægt (g/mol) 547.73
Formel vægt 547.353