missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Chymotrypsin-like protease Rabbit anti-Human, Mouse, Rat, Clone: 10Z4K3, Novus Biologicals™
Rabbit Monoclonal Antibody
1450.00 DKK - 3780.00 DKK
Tekniske data
| Antigen | Chymotrypsin-like protease |
|---|---|
| Klon | 10Z4K3 |
| Fortynding | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Applikationer | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
18310475
|
Novus Biologicals
NBP3-15845-20UL |
20 μg |
1450.00 DKK
20µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
18363705
|
Novus Biologicals
NBP3-15845-100UL |
100 μg |
3780.00 DKK
100µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
Chymotrypsin-like protease Monoclonal antibody specifically detects Chymotrypsin-like protease in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Tekniske data
| Chymotrypsin-like protease | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| chymotrypsin-like, chymotrypsin-like protease CTRL-1, CTRL1, EC 3.4.21, EC 3.4.21.-, MGC70821 | |
| A synthetic peptide corresponding to a sequence within amino acids 165-264 of human Chymotrypsin-like protease (P40313). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 10Z4K3 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1506 | |
| IgG | |
| Affinity purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel