missing translation for 'onlineSavingsMsg'
Learn More
Learn More
α-Chymotrypsin ≥40u/mg protein, MP Biochemicals™
Click to view available options
Quantity:
0.1 g
250 mg
500 mg
1 g
Specifications
Color | White |
Quantity | 250 mg |
Physical Form | Powder |
Chemical Name or Material | α-Chymotrypsin, 3X Crystallized from Bovine Pancreas, ≥40μ/mg protein |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'