missing translation for 'onlineSavingsMsg'
Learn More

Chymostatin MP Biomedicals

Product Code. 11434001 Shop All MP Biomedicals Products
Change view
Click to view available options
Mængde:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
11434001 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11434001 Supplier MP Biomedicals Supplier No. 0215284501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale Chymostatin
CAS 9076-44-2
Molekylær formel C31H41N7O6
Mængde 1 mg
Synonym chymostatin
InChI nøgle MRXDGVXSWIXTQL-MWVOZXJSSA-N
SMIL CC(C)CC(C(=O)NC(CC1=CC=CC=C1)C=O)NC(=O)C(C2CCN=C(N2)N)NC(=O)NC(CC3=CC=CC=C3)C(=O)O
IUPAC navn (2S)-2-[[(1S)-1-(2-amino-1,4,5,6-tetrahydropyrimidin-6-yl)-2-[[4-methyl-1-oxo-1-[[(2S)-1-oxo-3-phenylpropan-2-yl]amino]pentan-2-yl]amino]-2-oxo-yl]amino]-2-oxoylsyre
Molekylvægt (g/mol) 607.712
PubChem CID 129894295
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.