missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Chymase/CMA1/Mast Cell Chymase Antibody [HRP], Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals AF4099H
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Chymase/CMA1/Mast Cell Chymase Polyclonal antibody specifically detects Chymase/CMA1/Mast Cell Chymase in Human samples. It is validated for Immunoprecipitation,Western Blot
Tekniske data
| Chymase/CMA1/Mast Cell Chymase | |
| Polyclonal | |
| PBS | |
| Sheep | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| HRP | |
| Alpha-chymase, chymase, chymase 1 preproprotein transcript E, chymase 1 preproprotein transcript I, chymase 1, mast cell, chymase, heart, chymase, mast cell, CYH, CYM, EC 3.4.21, EC 3.4.21.39, Mast cell protease I, MCT1, MGC119890, MGC119891 | |
| Mouse myeloma cell line NS0-derived recombinant human Chymase/CMA1, Gly20-Asn247, Accession # P23946 | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Immunology, Innate Immunity, Mast Cell Markers | |
| 1215 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion