Learn More
Proteintech ChromoTek pSpot7 Vector
Product Code. 17323333
Change view
Click to view available options
Quantity:
1.25 μg
Unit Size:
1.25µg
Product Code. Quantity Unit Size
17323333 1.25 μg 1.25µg
1 options
Product Code. 17323333 Supplier Proteintech Supplier No. ev7
 Restricted Item
På lager
pSpot7 vector, S.cerevisiae, Spot N-term, Leu/Amp (Ori: 2 μ/pBR322), high expression.
  • The plasmid pSpot7 is an expression vector for Spot-Tag™ fusion proteins in S. cerevisiae. After cloning the protein of interest (POI) into the multiple cloning site (MCS) provided by pSpot7, the Spot-Tag™ (sequence: PDRVRAVSHWSS) will be fused to the N-terminus of the POI.
Specifications
Quantity 1.25 μg