Learn More
Macherey-Nagel™ Chromofil™ Glass Fiber Syringe Filter
Brand:  Macherey-Nagel™ 729034
Additional Details : Weight : 0.01000kg
Product Code. 10539985
  • 965.00 DKK / Pack of 100
Estimated Shipment: 10-06-2024
to see stock.
Specifications
Blue Top, Colorless Bottom
1 μm
100 Pack
15 mm
Glass Fiber
Solutions with High Loads of Particulate Matter
Syringe Filter