Få mere at vide
Macherey-Nagel™ Chrombond™ 150/50 SiOH Polypropylene Flash Column
For use with vacuum manifolds
Brand:  Macherey-Nagel™ 730887
Produktkode 12870631
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
  • Automated systems or CHROMABOND™ vacuum manifolds
  • Polypropylene column, polyethylene filter element
  • High loading capacity and separation efficiency
  • Constant back pressure, airtight sealing
  • Standard Luer port For automated solid phase extraction, parallel flash purification/synthesis, high throughput screening/purification.
Tekniske data
Tekniske data
170 mm
Flash Column
Biotage FlashMaster Systems
Polypropylene
Flash Column
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Korrektion af produktindhold
Produkttitel
Macherey-Nagel™ Chrombond™ 150/50 SiOH Polypropylene Flash Column > I.D.: 38.2mm