Få mere at vide
Purolite™ Chromalite™ AD Monodispersed Polymeric Adsorbents for IEX and RP-HPLC
Produkttype:
10AD2 Makroporøs polystyren/divinylbenzen-adsorbent
10AD2Q Makroporøs polystyren/divinylbenzen anionbytterharpiks
10AD2S Makroporøs polystyren/divinylbenzen kationbytterharpiks
15AD2 Makroporøs polystyren/divinylbenzen-adsorbent
15AD2Q Makroporøs polystyren/divinylbenzen anionbytterharpiks
15AD2S Makroporøs polystyren/divinylbenzen kationbytterharpiks
30AD2 Makroporøs polystyren/divinylbenzen-adsorbent
5AD2 Makroporøs polystyren/divinylbenzen-adsorbent
Emballagemængde:
10 g
50 g
100 g
Pakningsstørrelse:
10g
50g
Beskrivelse
- Manufactured using patented seeding technology foir uniform particle sizes
- Solid Phase Extraction
- Analytical Reverse Phase chromatography (RP-HPLC) and IEX chromatography (IEX-HPLC)
- Adsorption of biomolecules
- Extraction from biological fluid
- High chemical stability
- Efficient regeneration
- Large surface area
- Narrow particle size range
- Excellent selectivity
- Low extractable content
- Compliant with FDA regulation
- Compliant with ResAP(2004) 3
- Halal, Kosher, TSE/BSE/GMO free
Tekniske data
Tekniske data
| Form | White to Off-White Spherical Beads |
| Opbevaringskrav | 2°C to 20°C |
| Produkttype | 15AD2 Makroporøs polystyren/divinylbenzen-adsorbent |
| Partikelstørrelse | 12 to 18 μm |
| pH-område | 1 to 14 pH |
| Porestørrelse | 200 to 300 Å |
| Overfladeareal (metrisk) | >500 m2 |
| Emballagemængde | 10 g |
| Indhold og opbevaring | Use within 5 years from date of manufacture |
| Format | 20% ethanol slurry |