Learn More
Macherey-Nagel™ Chromafil™ Multi 96 Filter Plates
Brand: Macherey-Nagel™ 738771.M
Additional Details : Weight : 0.01000kg
Specifications
Standard | |
0.45 μm | |
96 |
8 x 12 | |
Chromafil Multi 96 Filter Plates | |
With cellulose mixed ester filter elements |