Learn More
Macherey-Nagel™ Chromafil™ Multi 96 Filter Plates
Brand:  Macherey-Nagel™ 738771.M
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12451778
  • 270.00 DKK / Each
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
Specifications
Specifications
Standard
0.45 μm
96
8 x 12
Chromafil Multi 96 Filter Plates
With cellulose mixed ester filter elements