Learn More
Thermo Scientific™ Chromacol 8mm Screw Caps
Thermo Scientific™ Chromacol Screw Caps are offered with a selection of liners and colors.
Brand: Thermo Scientific™ 8-SC-ST14
Additional Details : Weight : 0.28000kg
Description
Thermo Scientific™ Chromacol Screw Caps are offered with a selection of liners and colors.
Specifications
8mm Screw Cap; Blue; Prefitted Silicone/PTFE Liner | |
Blue | |
Chromacol |
Silicone/PTFE | |
Prefitted Silicone |