Learn More
Macherey-Nagel™ Chromabond™ Drug Polypropylene SPE Column
Brand: Macherey-Nagel™ 730682
Additional Details : Weight : 0.20000kg
Specifications
Silica | |
SPE | |
SPE Enrichment of Drugs From Urine or Plasma | |
Chromabond Drug | |
60 Å |
6 mL | |
500 mg | |
45 μm | |
Solid Phase Extraction Column | |
500 m2/g |