missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chordin Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99454G
Additional Details : Weight : 0.00970kg
Description
Chordin Polyclonal specifically detects Chordin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Chordin | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CHRD, MGC133038 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Chordin. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
8646 | |
Store at 4°C in the dark. | |
IgG |