Få mere at vide
Choline-d9 Chloride, TRC
Artikelnummer. 30364547
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30364547 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30364547 Leverandør Toronto Research Chemicals Leverandørnr. TRCC43265210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Choline-D9 Chloride
CAS 61037-86-3
Renhedsgrad noter HPLC
Molekylær formel C5 H5 D9 N O . Cl
Mængde 10 mg
Synonym 2-Hydroxy-N,N,N-trimethylethanaminium-d9 Chloride, (Trimethyl-d9)(2-hydroxyethyl)ammonium Chloride, Actiron CC 6-d9, Bilineurin-d9 Chloride, Biocolina-d9, Hepacholine-d9, Lipotril-d9, Neocolina-d9, Paresan-d9
InChI formel InChI=1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1/i1D3,2D3,3D3;
SMIL C([N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])CO.[Cl-]
IUPAC navn 2-hydroxy-N,N,N-tris(methyl-d3)ethan-1-aminium chloride
Molekylvægt (g/mol) 148.68
Formel vægt 148.13
Procent renhed >95