Få mere at vide
Choline-d13 Chloride (N,N,N-trimethyl-d9; 1,1,2,2-d4), TRC
Artikelnummer. 30352304
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30352304 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30352304 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4326511MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Choline-d13 Chloride (N,N,N-trimethyl-d9; 1,1,2,2-d4)
CAS 352438-97-2
Molekylær formel C5HD13ClNO
Mængde 1 mg
Synonym Ethan-1,1,2,2-d4-aminium, 2-hydroxy-N,N,N-tri(methyl-d3)-, chloride (9CI), Choline-d13 Chloride, Choline-D13 Chloride, 2-Hydroxy-N,N,N-tri(methyl-d3)ethan-1,1,2,2-d4-aminium chloride (1:1), Choline-d13 Chloride (N,N,N-trimethyl-d9, 1,1,2,2-d4)
InChI formel InChI=1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1/i1D3,2D3,3D3,4D2,5D2;
SMIL C([N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])(C(O)([2H])[2H])([2H])[2H].[Cl-]
IUPAC navn 2-hydroxy-N,N,N-tris(methyl-d3)ethan-1-aminium-1,1,2,2-d4 chloride
Molekylvægt (g/mol) 152.7
Formel vægt 152.16