Få mere at vide
Choline-d13 Bromide (N,N,N-trimethyl-d9; 1,1,2,2-d4), CDN
Artikelnummer. 30322067
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322067 0.25 g -
30312203 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322067 Leverandør CDN Leverandørnr. CDND03850.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Choline-D13 Bromide
CAS 203645-64-1
Molekylær formel C5 D13 H N O . Br
Mængde 0.25 g
Synonym Ethan-1,1,2,2-d4-aminium, 2-hydroxy-N,N,N-tri(methyl-d3)-, bromide (9 CI), Choline-d13 Bromide, Choline-D13 Bromide, 2-Hydroxy-N,N,N-tri(methyl-d3)ethan-1,1,2,2-d4-aminium bromide (1:1), Choline-d13 Bromide (N,N,N-trimethyl-d9, 1,1,2,2-d4)
InChI formel InChI=1 S/C5H14NO.BrH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1 H/q+1;/p-1/i1D3,2D3,3D3,4D2,5D2;
SMIL [Br-].[2 H]C([2 H])([2 H])[N+](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])O
IUPAC navn (1,1,2,2-tetradeuterio-2-hydroxyethyl)-tris(trideuteriomethyl)azanium;bromide
Molekylvægt (g/mol) 197.1549
Formel vægt 196.1075 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity