missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cholic Acid Sodium Salt, ≥97% (dry basis), Ultrapure, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
Packaging:
Plastic bottle
Specifications
Physical Form | Powder |
Quantity | 100 g |
Beilstein | 3582354 |
Formula Weight | 430.55 |
Percent Purity | ≥98% |
Grade | Ultrapure |
Packaging | Plastic bottle |
Assay | Dry basis |
Chemical Name or Material | Cholic Acid Sodium Salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'