Få mere at vide
Cholic Acid-d5, TRC
Artikelnummer. 30366693
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30366693 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366693 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4326021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cholic Acid-d5
CAS 53007-09-3
Renhedsgrad noter HPLC
Molekylær formel C24H35D5O5
Mængde 1 mg
Synonym (3α,5β,7α12α)-3,7,12-Trihydroxy-cholan-24-oic Acid-d5, 17β-(1-Methyl-3-carboxy-propyl)etiocholane-3α,7α,12α-triol-d5, 5β-Cholic Acid-d5, Cholalic Acid-d5, Cholalin-d5, Colalin-d5, NSC-6135-d5
InChI formel InChI=1S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29)/t13-,14+,15-,16-,17+,18?,19-,20+,22?,23+,24-/m1/s1/i8D2,10D2,15D
SMIL O[C@]1([2H])C([2H])([2H])C[C@@]2(C)[C@@](C[C@@H](O)C3C2C[C@H](O)[C@@]4(C)[C@@]3([H])CC[C@@H]4[C@@H](CCC(O)=O)C)([H])C1([2H])[2H]
IUPAC navn (4R)-4-[(3R,5S,7R,10S,12S,13R,14S,17R)-2,2,3,4,4-pentadeuterio-3,7,12-trihydroxy-10,13-dimethyl-1,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]pentanoic acid
Molekylvægt (g/mol) 413.6
Procent renhed >95