Få mere at vide
Cholic Acid-d4, TRC
Artikelnummer. 30352303
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30352303 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30352303 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4326035MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cholic Acid-d4
CAS 116380-66-6
Renhedsgrad noter HPLC
Molekylær formel C24 D4 H36 O5
Mængde 5 mg
Synonym Cholan-24-oic-2,2,4,4-d4 acid, 3,7,12-trihydroxy-, (3α,5β,7α,12α)- (9CI, ACI), (3α,5β,7α,12α)-3,7,12-Trihydroxycholan-24-oic-2,2,4,4-d4 acid (ACI), Cholic acid-2,2,4,4-d4, Cholic acid-d4, Cholic-2,2,4,4-d4 Acid, Cholic Acid-2,2,4,4-D4
InChI formel InChI=1S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29)/t13-,14+,15-,16-,17+,18+,19-,20+,22+,23+,24-/m1/s1/i8D2,10D2
SMIL [2H]C1([2H])C[C@]2(C)[C@H]3C[C@H](O)[C@]4(C)[C@H](CC[C@H]4[C@@H]3[C@H](O)C[C@@H]2C([2H])([2H])[C@@H]1O)[C@H](C)CCC(=O)O
IUPAC navn (4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-2,2,4,4-tetradeuterio-3,7,12-trihydroxy-10,13-dimethyl-3,5,6,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molekylvægt (g/mol) 412.596
Formel vægt 412.313
Procent renhed >95