missing translation for 'onlineSavingsMsg'
Learn More

Cholic Acid Acyl Glucuronide, TRC

Product Code. 30135954
Change view
Click to view available options
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30135954 25 mg
30011594 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135954 Supplier Toronto Research Chemicals Supplier No. TRCA30535025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208038-30-6
Molekylær formel C30 H48 O11
Molekylvægt (g/mol) 584.696
Synonym (2S,3S,4S,5R,6S)-3,4,5-Trihydroxy-6-(((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoyl)oxy)tetrahydro-2H-pyran-2-carboxylic Acid, β-D-Glucopyranuronic Acid, 1-[(3α,5β,7α,12α)-3,7,12-Trihydroxycholan-24-oate;
IUPAC navn (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]oxyoxane-2-carboxylic acid
SMIL C[C@H](CCC(=O)O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)O)[C@H]2CC[C@H]3[C@@H]4[C@H](O)C[C@@H]5C[C@H](O)CC[C@]5(C)[C@H]4C[C@H](O)[C@]23C

Specifications

Kemisk navn eller materiale Cholic Acid Acyl Glucuronide
Mængde 25 mg
InChI formel InChI=1S/C30H48O11/c1-13(4-7-21(34)40-28-25(37)23(35)24(36)26(41-28)27(38)39)16-5-6-17-22-18(12-20(33)30(16,17)3)29(2)9-8-15(31)10-14(29)11-19(22)32/h13-20,22-26,28,31-33,35-37H,4-12H2,1-3H3,(H,38,39)/t13-,14+,15-,16-,17+,18+,19-,20+,22+,23+,24+,25-,26+,28-,29+,30-/m1/s1
Formel vægt 584.32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.