Få mere at vide
Cholesteryl Hemisuccinate, TRC
Artikelnummer. 30366312
Skift visning
Mængde:
1 g
2.5 g
5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30366312 2.5 g 2.5g
30349364 5 g -
30360007 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30366312 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4325862.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1510-21-0
Molekylær formel C31H50O4
Molekylvægt (g/mol) 486.73
Synonym 3-(3-Cholesteryloxycarbonyl)propionic Acid, 3-Cholesteryloxycarbonylpropanoic Acid, 3-[(Cholest-5-en-3β-yl)oxycarbonyl]propionic Acid, Cholesterol Hemisuccinate, Cholesterol Monosuccinate, Cholesteryl Hemisuccinate, Cholesteryl Hydrogen Succinate, Cholesteryl Monosuccinate, Mono(cholesteryl) Succinate, Succinate Cholesterol Monoester, Succinic Acid Monocholesteryl Ester
IUPAC navn 4-[[(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl]oxy]-4-oxobutanoic acid
SMIL CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(=O)CCC(=O)O
Tekniske data
Kemisk navn eller materiale Cholesteryl Hemisuccinate
Mængde 2.5 g
InChI formel InChI=1S/C31H50O4/c1-20(2)7-6-8-21(3)25-11-12-26-24-10-9-22-19-23(35-29(34)14-13-28(32)33)15-17-30(22,4)27(24)16-18-31(25,26)5/h9,20-21,23-27H,6-8,10-19H2,1-5H3,(H,32,33)/t21-,23+,24+,25-,26+,27+,30+,31-/m1/s1