Få mere at vide
Cholesteryl Decanoate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Cholesteryl Decanoate |
| CAS | 1183-04-6 |
| Molekylær formel | C37H64O2 |
| Mængde | 25 g |
| Synonym | (3beta)-Cholest-5-en-3-ol Decanoate, Cholesteryl Ester Decanoic Acid, 3beta-(Decanoyloxy)-5-cholestene, 5-Cholesten-3beta-ol Caprate, Cholest-5-en-3beta-ol Caprate, Cholesterol 3-Decanoate, Cholesterol Caprate, Cholesterol n-Decylate, Cholesteryl Caprate, Cholesteryl Caprinate, Cholesteryl Decanoate, Cholesteryl Decylate, Cholesteryl n-Decylate |
| InChI formel | InChI=1S/C37H64O2/c1-7-8-9-10-11-12-13-17-35(38)39-30-22-24-36(5)29(26-30)18-19-31-33-21-20-32(28(4)16-14-15-27(2)3)37(33,6)25-23-34(31)36/h18,27-28,30-34H,7-17,19-26H2,1-6H3/t28-,30+,31+,32-,33+,34+,36+,37-/m1/s1 |
| SMIL | CCCCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C |
| IUPAC navn | [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] decanoate |
| Molekylvægt (g/mol) | 540.9 |
| Formel vægt | 540.4906 |