Få mere at vide
Cholesteryl-2,2,3,4,4,6-d6 Octadecanoate, CDN
Artikelnummer. 30323585
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323585 0.05 g
30311859 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323585 Leverandør CDN Leverandørnr. CDND58230.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cholesteryl-2,2,3,4,4,6-D6 Stearate
CAS 2692624-28-3
Molekylær formel C45 D6 H74 O2
Mængde 0.05 g
Synonym Cholest-5-en-2,2,3,4,4,6-d6-3-ol, 3-octadecanoate, (3β)- (ACI), (3β)-Cholest-5-en-2,2,3,4,4,6-d6-3-ol 3-octadecanoate, Cholesteryl-2,2,3,4,4,6-d6 Octadecanoate, Cholesteryl-2,2,3,4,4,6-D6 Stearate, Cholesterol stearate-d6, Stearic acid cholesteryl-2,2,3,4,4,6-d6 ester
InChI formel InChI=1 S/C45H80O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-25-43(46)47-38-30-32-44(5)37(34-38)26-27-39-41-29-28-40(36(4)24-22-23-35(2)3)45(41,6)33-31-42(39)44/h26,35-36,38-42 H,7-25,27-34H2,1-6H3/t36-,38+,39+,40-,41+,42+,44+,45-/m1/s1/i26D,30D2,34D2,38 D
SMIL [2 H]C1=C2C([2 H])([2 H])[C@@]([2 H])(OC(=O)CCCCCCCCCCCCCCCCC)C([2 H])([2 H])C[C@]2(C)[C@H]3 CC[C@]4(C)[C@H](CC[C@H]4 [C@@H]3C1)[C@H](C)CCCC(C)C
IUPAC navn [(3 S,8 S,9 S,10 R,13 R,14 S,17 R)-2,2,3,4,4,6-hexadeuterio-10,13-dimethyl-17-[(2 R)-6-methylheptan-2-yl]-1,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl] octadecanoate
Molekylvægt (g/mol) 659.1525
Formel vægt 658.6535 g/mol
Procent renhed 98 atom % D, min 97% Chemical Purity