Få mere at vide
Cholesterol-d7, TRC
Artikelnummer. 30367352
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30367352 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367352 Leverandør Toronto Research Chemicals Leverandørnr. TRCC432503100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cholesterol-d7
CAS 83199-47-7
Molekylær formel C27 2H7 H39 O
Mængde 100 mg
Synonym Cholest-5-en-25,26,26,26,27,27,27-d7-3-ol, (3β)- (9CI, ACI), (3β)-Cholest-5-en-25,26,26,26,27,27,27-d7-3-ol (ACI), [25,26,26,26,27,27,27-d7]-Cholesterol, Cholesterol-25,26,26,26,27,27,27-d7, Cholesterol-d7, Cholesterol-25,26,26,26,27,27,27-D7
InChI formel InChI=1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i1D3,2D3,18D
SMIL [2H]C([2H])([2H])C([2H])(CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C([2H])([2H])[2H]
IUPAC navn (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6,7,7,7-tetradeuterio-6-(trideuteriomethyl)heptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
Molekylvægt (g/mol) 393.7
Formel vægt 393.3988