Få mere at vide
Cholesterol-d6, TRC
Artikelnummer. 30353020
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30353020 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353020 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4325022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cholesterol-d6
CAS 92543-08-3
Molekylær formel C27 2H6 H40 O
Mængde 2.5 mg
Synonym Cholesterol-2,2,3,4,4,6-d6, (3β)-Cholest-5-en-3-ol-d6, (3β)-Cholest-5-en-3-ol-d6, (-)-Cholesterol-d6, 3β-Hydroxycholest-5-ene-d6, 5:6-Cholesten-3β-ol-d6, Cholest-5-en-3β-ol-d6, Cholesterin-d6, Cholesteryl-d6 αlcohol, Dythol-d6, Lidinit-d6, Lidinite-d6, NSC 8798-d6, Provitamin D-d6, 5-Cholesten-3β-ol-d6
InChI formel InChI=1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i9D,13D2,17D2,21D
SMIL [2H]C1=C2C([2H])([2H])[C@@]([2H])(O)C([2H])([2H])C[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3C1)[C@H](C)CCCC(C)C
IUPAC navn (3S,8S,9S,10R,13R,14S,17R)-2,2,3,4,4,6-hexadeuterio-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-ol
Molekylvægt (g/mol) 392.69
Formel vægt 392.3925