Få mere at vide
Cholesterol-d4, TRC
Artikelnummer. 30360064
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30360064 2.5 mg 2.5mg
30361950 10 mg -
30368414 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30360064 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4325042.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 956029-28-0
Molekylær formel C27 2H4 H42 O
Molekylvægt (g/mol) 390.68
Synonym Cholest-5-en-25,26,26,26-d4-3-ol, (3β)- (ACI), (3β)-Cholest-5-en-25,26,26,26-d4-3-ol (ACI), [25,26,26,26-2H4]-Cholesterol, Cholesterol-25,26,26,26-d4, Cholesterol-d4, Cholesterol-25,26,26,26-D4
IUPAC navn (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6,7,7,7-tetradeuterio-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMIL [2H]C([2H])([2H])C([2H])(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale Cholesterol 6,7,7-d4
Mængde 2.5 mg
InChI formel InChI=1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i1D3,18D/t18?,19-,21+,22+,23-,24+,25+,26+,27-
Formel vægt 390.38