Få mere at vide
Cholesterol-3-d1, CDN
Artikelnummer. 30315741
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315741 0.005 g
30320809 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315741 Leverandør CDN Leverandørnr. CDND12200.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cholesterol-3-D
CAS 51467-57-3
Molekylær formel C27 D H45 O
Mængde 0.005 g
Synonym Cholest-5-en-3-d-3-ol, (3β)- (9 CI, ACI), (3β)-Cholest-5-en-3-d-3-ol (ACI), (3β)-Cholest-5-en-3-ol-3 d, [3-2H1]-Cholesterol, Cholesterol-3-d, Cholesterol-3-D, Cholesterol-d1
InChI formel InChI=1 S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28 H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i21D
SMIL [2 H][C@]1(O)CC[C@]2(C)[C@H]3 CC[C@]4(C)[C@H](CC[C@H]4 [C@@H]3 CC=C2C1)[C@H](C)CCCC(C)C
IUPAC navn (3 S,8 S,9 S,10 R,13 R,14 S,17 R)-3-deuterio-10,13-dimethyl-17-[(2 R)-6-methylheptan-2-yl]-1,2,4,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-ol
Molekylvægt (g/mol) 387.6597
Formel vægt 387.3611 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity