Få mere at vide
Cholesterol 3-Acetate-d7, TRC
Artikelnummer. 30366514
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366514 25 mg
30364069 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366514 Leverandør Toronto Research Chemicals Leverandørnr. TRCC43250725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1345629-08-4
Molekylær formel C29H41D7O2
Molekylvægt (g/mol) 435.73
Synonym (-)-Cholesteryl Acetate-d7, (3β)-Cholest-5-en-3-ol Acetate-d7, 3-Cholesteryl Acetate-d7, 3β-Acetoxycholest-5-ene-d7, Cholest-5-en-3β-ol Acetate-d7, Cholest-5-en-3β-yl Acetate-d7, Cholesterin Acetate-d7, Cholesterol 3-Acetate-d7, Cholesterol 3β-Acetate-d7, Cholesteryl Acetate-d7, NSC 8799
IUPAC navn [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6,7,7,7-tetradeuterio-6-(trideuteriomethyl)heptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
SMIL [H][C@@]12CC=C3C[C@@H](OC(C)=O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4([H])[C@H](C)CCCC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale Cholesterol 3-Acetate-d7
Mængde 25 mg
InChI formel InChI=1S/C29H48O2/c1-19(2)8-7-9-20(3)25-12-13-26-24-11-10-22-18-23(31-21(4)30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-20,23-27H,7-9,11-18H2,1-6H3/t20-,23+,24+,25-,26+,27+,28+,29-/m1/s1/i1D3,2D3,19D