Få mere at vide
Cholesta-4,6-dien-3-one, TRC
Artikelnummer. 30349274
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30349274 2.5 mg
30351040 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30349274 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4314202.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 566-93-8
Molekylær formel C27 H42 O
Molekylvægt (g/mol) 382.62
Synonym 4,6-Cholestadien-3-one, NSC 119073
IUPAC navn (8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-one
SMIL CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3C=CC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale Cholesta-4,6-dien-3-one
Mængde 2.5 mg
InChI formel InChI=1S/C27H42O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9-10,17-19,22-25H,6-8,11-16H2,1-5H3/t19-,22+,23-,24+,25+,26+,27-/m1/s1
Formel vægt 382.3236