Få mere at vide
Chlorzoxazone-4,6,7-d3, TRC
Artikelnummer. 17998209
Skift visning
Mængde:
1 mg
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17998209 1 mg -
17987939 10 mg -
17909229 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 17998209 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4287021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1185173-60-7
Molekylær formel C7 D3 H Cl N O2
Molekylvægt (g/mol) 172.584
Synonym 5-Chloro-4,6,7-trideuterio-1,3-benzoxazol-2-ol, 5-Chloro-1,3-benzoxazol-2(3H)-one-4,6,7-d3, 5-Chloro-2-benzoxazolinone-4,6,7-d3, Chlorzoxazone-d3, Chlorzoxazone-4,6,7-D3
IUPAC navn 5-chloro-4,6,7-trideuterio-3H-1,3-benzoxazol-2-one
SMIL [2H]c1c([2H])c2OC(=O)Nc2c([2H])c1Cl
Tekniske data
Kemisk navn eller materiale Chlorzoxazone-4,6,7-d3
Mængde 1 mg
InChI formel InChI=1S/C7H4ClNO2/c8-4-1-2-6-5(3-4)9-7(10)11-6/h1-3H,(H,9,10)/i1D,2D,3D
Formel vægt 172.012