Få mere at vide
Chlortoluron-d6 (N,N-dimethyl-d6), CDN
Artikelnummer. 30315654
Skift visning
Mængde:
0.05 g
Artikelnummer. Mængde
30315654 0.05 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315654 Leverandør CDN Leverandørnr. CDND71280.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlorotoluron (N,N-dimethyl) D6
CAS 1219803-48-1
Molekylær formel C10 2H6 H7 Cl N2 O
Mængde 0.05 g
Synonym 3-(3-Chloro-4-methylphenyl)-1,1-bis(trideuteriomethyl)urea, Chlorotoluron D6 (N,N-dimethyl D6)
InChI formel InChI=1 S/C10H13ClN2O/c1-7-4-5-8(6-9(7)11)12-10(14)13(2)3/h4-6 H,1-3H3,(H,12,14)/i2D3,3D3
SMIL [2 H]C([2 H])([2 H])N(C(=O)Nc1ccc(C)c(Cl)c1)C([2 H])([2 H])[2 H]
IUPAC navn 3-(3-chloro-4-methylphenyl)-1,1-bis(trideuteriomethyl)urea
Molekylvægt (g/mol) 218.71
Formel vægt 218.1093 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity