Få mere at vide
Chlorthiamid, TRC
Artikelnummer. 30352985
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30352985 100 mg
30365540 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30352985 Leverandør Toronto Research Chemicals Leverandørnr. TRCC369943100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1918-13-4
Molekylær formel C7 H5 Cl2 N S
Molekylvægt (g/mol) 206.09
Synonym Benzamide, 2,6-dichlorothio- (7CI,8CI), 2,6-Dichlorobenzenecarbothioamide, 2,6-Dichlorothiobenzamide, Chlorthiamid, Chlorthiamide, DCBN, Prefix, Prefix (chlorthiamid), SD 7961
IUPAC navn 2,6-dichlorobenzenecarbothioamide
SMIL NC(=S)c1c(Cl)cccc1Cl
Tekniske data
Kemisk navn eller materiale Chlorthiamid
Mængde 100 mg
InChI formel InChI=1S/C7H5Cl2NS/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H2,10,11)
Formel vægt 204.952