missing translation for 'onlineSavingsMsg'
Få mere at vide

Chlorthalidone-d4, TRC

Artikelnummer. 30365352
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365352 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365352 Leverandør Toronto Research Chemicals Leverandørnr. TRCC42750225MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Chlortalidone isoindol-4,5,6,7-d4
CAS 1794941-44-8
Renhedsgrad noter HPLC
Molekylær formel C14 2H4 H7 Cl N2 O4 S
Mængde 25 mg
Synonym 2-Chloro-5-(4,5,6,7-tetradeuterio-1-hydroxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide, Chlorthalidone-d4, Chlortalidone-D4 (isoindol-4,5,6,7-D4), 2-Chloro-5-(1-hydroxy-3-oxo-1-isoindolinyl-4,5,6,7-d4)benzenesulfonamide, 3-Hydroxy-3-[4-chloro-3-sulfamylphenyl]phthalimidine-4,5,6,7-d4
InChI formel InChI=1S/C14H11ClN2O4S/c15-11-6-5-8(7-12(11)22(16,20)21)14(19)10-4-2-1-3-9(10)13(18)17-14/h1-7,19H,(H,17,18)(H2,16,20,21)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c2c(C(=O)NC2(O)c3ccc(Cl)c(c3)S(=O)(=O)N)c1[2H]
IUPAC navn 2-chloro-5-(4,5,6,7-tetradeuterio-1-hydroxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide
Molekylvægt (g/mol) 342.79
Formel vægt 342.0379
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.