Få mere at vide
Chlorthalidone-d4, TRC
Artikelnummer. 30350332
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30350332 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350332 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4275021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlortalidone isoindol-4,5,6,7-d4
CAS 1794941-44-8
Renhedsgrad noter HPLC
Molekylær formel C14 2H4 H7 Cl N2 O4 S
Mængde 1 mg
Synonym 2-Chloro-5-(4,5,6,7-tetradeuterio-1-hydroxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide, Chlorthalidone-d4, Chlortalidone-D4 (isoindol-4,5,6,7-D4), 2-Chloro-5-(1-hydroxy-3-oxo-1-isoindolinyl-4,5,6,7-d4)benzenesulfonamide, 3-Hydroxy-3-[4-chloro-3-sulfamylphenyl]phthalimidine-4,5,6,7-d4
InChI formel InChI=1S/C14H11ClN2O4S/c15-11-6-5-8(7-12(11)22(16,20)21)14(19)10-4-2-1-3-9(10)13(18)17-14/h1-7,19H,(H,17,18)(H2,16,20,21)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c2c(C(=O)NC2(O)c3ccc(Cl)c(c3)S(=O)(=O)N)c1[2H]
IUPAC navn 2-chloro-5-(4,5,6,7-tetradeuterio-1-hydroxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide
Molekylvægt (g/mol) 342.79
Formel vægt 342.0379
Procent renhed >95